Drying Tubes, Straight, Top and Outer Joints, Chemglass
80067-982
Small Business Enterprise
- [ST] joint no.:14/20
- Description:Drying Tube, Straight
- Cat. No.:80067-982
- Supplier no.:CG-1294-14
Drying tubes feature straight, top and outer joints.
- Top standard taper outer joint
- Same size as the lower inner joint
Similar to CG-1293 but with the addition of a top standard taper outer joint of the same size as the lower inner joint.