Human Recombinant MCP-1 (from E. coli)
Catalog # 76303-830
Supplier: Peprotech
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Biological Activity:Determined by its ability to chemoattract human monocytes using a concentration range of 10.0-100.0 ng/ml.
- Gene ID:6347
- Animal-Free:Y
- Protein Synonyms:MCAF|Monocyte Chemotactic Protein-1|CCL2
- Protein/Peptide Name:MCP-1
- Purity:98%
- Molecular Weight:8.6 kDa
- Sequence:QPDAINAPVT CCYNFTNRKI SVQRLASYRR ITSSKCPKEA VIFKTIVAKE ICADPKQKWV QDSMDHLDKQ TQTPKT
- Endotoxin Level:Low
- Formulation:Lyophilized
- Shipping Temperature:Ambient
- Cat. No.:76303-830
Specifications
About this item
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Human MCP-1 is an 8.6 kDa protein containing 76 amino acid residues, including the four highly conserved cysteine residues present in the CC chemokines.