Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant FGF-BP-1 (from E. coli), PeproTech
 :  Peprotech
 
Human Recombinant FGF-BP-1 (from E. coli), PeproTech
 :  Peprotech

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological Activity:
    Determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors. The expected ED50 for this effect is 1.5-3.0 ug/ml.
  • Gene ID:
    9982
  • Animal-Free:
    Y
  • Protein Synonyms:
    FGF-binding protein 1|HBp17|FGF-BP|Fibroblast growth factor binding protein 1
  • Protein/Peptide Name:
    FGF-BP-1
  • Purity:
    95%
  • Molecular Weight:
    23.9 kDa
  • Sequence:
    MKKKVKNGLH SKVVSEQKDT LGNTQIKQKS RPGNKGKFVT KDQANCRWAA TEQEEGISLK VECTQLDHEF SCVFAGNPTS CLKLKDERVY WKQVARNLRS QKDICRYSKT AVKTRVCRKD FPESSLKLVS STLFGNTKPR KEKTEMSPRE HIKGKETTPS SLAVTQTMAT KAPECVEDPD MANQRKTALE FCGETWSSLC TFFLSIVQDT SC
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient

 

Frequently Bought Together