Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant FGF-4 (from E. coli)
Human Recombinant FGF-4 (from E. coli)
Catalog # 76303-684
Supplier:  Peprotech
CAS Number:  
Human Recombinant FGF-4 (from E. coli)
Catalog # 76303-684
Supplier:  Peprotech
Supplier Number:  100-31-50UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Biological Activity:
    Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 10 ng/ml, corresponding to a specific activity of ≥ 1 x 10^5 units/mg.
  • Gene ID:
    2249
  • Animal-Free:
    Y
  • Protein Synonyms:
    Transforming protein KS3|HST-1|HBGF-4|Fibroblast Growth Factor-4
  • Protein/Peptide Name:
    FGF-4
  • Purity:
    95%
  • Molecular Weight:
    19.7 kDa
  • Sequence:
    GRGGAAAPT APNGTLEAEL ERRWESLVAL SLARLPVAAQ PKEAAVQSGA GDYLLGIKRL RRLYCNVGIG FHLQALPDGR IGGAHADTRD SLLELSPVER GVVSIFGVAS RFFVAMSSKG KLYGSPFFTD ECTFKEILLP NNYNAYESYK YPGMFIALSK NGKTKKGNRV SPTMKVTHFL PRL
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    76303-684

Specifications

About this item

FGF-4 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-4 signals through FGFR 1c, 2c, 3c, and 4. Recombinant Human FGF-4 is a 19.7 kDa protein consisting of 182 amino acid residues.