About this item
IL-21 is a pleiotropic cytokine produced by CD4+ T cells in response to antigenic stimulation. Its action generally enhances antigen-specific responses of immune cells. The biological effects of IL-21 include: inducing the differentiation of T cell-stimulated B cells into plasma cells and memory B cells; the stimulation of IgG production in conjunction with IL-4; and the induction of apoptotic effects in naive B cells and stimulated B cells in the absence of T-cell signaling. Additionally, IL-21 promotes the anti-tumor activity of CD8+ T cells and NK cells. IL-21 exerts its effect through binding to a specific type I cytokine receptor, IL-21R, which also contains the gamma chain (gammac) found in other cytokine receptors, including IL-2, IL-4, IL-7, IL-9 and IL-15. The IL-21/IL-21R interaction triggers a cascade of events, which includes activation of the tyrosine kinases JAK1 and JAK3, followed by activation of the transcription factors STAT1 and STAT3. Recombinant Murine IL-21 is a 15.0 kDa protein consisting of 130 amino acid residues.
- Animal-free
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Biological Activity:Determined by its ability to stimulate the proliferation of human ANBL-6 cells. The expected ED50 is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg.
- Gene ID:60505
- Animal-Free:Y
- Protein Synonyms:Za11|Interleukin-21
- Protein/Peptide Name:IL-21
- Purity:98%
- Molecular Weight:15 kDa
- Sequence:MHKSSPQGPD RLLIRLRHLI DIVEQLKIYE NDLDPELLSA PQDVKGHCEH AAFACFQKAK LKPSNPGNNK TFIIDLVAQL RRRLPARRGG KKQKHIAKCP SCDSYEKRTP KEFLERLKWL LQKMIHQHLS
- Endotoxin Level:Low
- Formulation:Lyophilized
- Shipping Temperature:Ambient