Human Recombinant TECK (CCL25) (from E. coli)
Supplier: Peprotech
Certificates
About this item
TECK is a CC chemokine, specifically expressed by thymic stromal cells, and signals through the CCR9 receptor. TECK is chemotactic towards activated macrophages, thymocytes and dendritic cells. Recombinant Human TECK (CCL25) is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
- Animal-free
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Biological Activity:Not available.
- Gene ID:6370
- Animal-Free:Y
- Protein Synonyms:Thymus Expressed Chemokine
- Protein/Peptide Name:TECK (CCL25)
- Purity:98%
- Molecular Weight:14.2 kDa
- Sequence:QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN MQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSKRNVSLL ISANSGL
- Endotoxin Level:Low
- Formulation:Lyophilized
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together