Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant sRANK Ligand (from E. coli)
Human Recombinant sRANK Ligand (from E. coli)
Catalog # 76303-648
Supplier:  Peprotech
CAS Number:  
Human Recombinant sRANK Ligand (from E. coli)
Catalog # 76303-648
Supplier:  Peprotech
Supplier Number:  AF-310-01-50UG
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Biological Activity:
    Determined by its dose-dependent ability to induce reporter gene in HT-29 NF-κB Luc reporter cells.
  • Gene ID:
    8600
  • Animal-Free:
    Y
  • Protein Synonyms:
    TRANCE (TNF-related activation-induced cytokine)|OPGL|TNFSF11|ODF (Osteoclast differentiation factor)|soluble Receptor Activator of NF-κB Ligand
  • Protein/Peptide Name:
    sRANK Ligand
  • Purity:
    98%
  • Molecular Weight:
    20 kDa
  • Sequence:
    MEKAMVDGSW LDLAKRSKLE AQPFAHLTIN ATDIPSGSHK VSLSSWYHDR GWAKISNMTF SNGKLIVNQD GFYYLYANIC FRHHETSGDL ATEYLQLMVY VTKTSIKIPS SHTLMKGGST KYWSGNSEFH FYSINVGGFF KLRSGEEISI EVSNPSLLDP DQDATYFGAF KVRDID
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    76303-648

Specifications

About this item

RANKL and RANK are members of the TNF superfamily of ligands and receptors that play an important role in the regulation of specific immunity and bone turnover. RANK (receptor) was originally identified as a dendritic cell-membrane protein, which, by interacting with RANKL, augments the ability of dendritic cells. These dendritic cells then stimulate naive T-cell proliferation in a mixed lymphocyte reaction, promote the survival of RANK+ T-cells, and regulate T-cell-dependent immune response. RANKL, which is expressed in a variety of cells, including osteoblasts, fibroblasts, activated T-cells and bone marrow stromal cells, is also capable of interacting with a decoy receptor called OPG. Binding of soluble OPG to sRANKL inhibits osteoclastogenesis by interrupting the signaling between stromal cells and osteoclastic progenitor cells, thereby leading to excess accumulation of bone and cartilage. Human RANKL is reactive on murine cells. Recombinant Human sRANK Ligand is a 20.0 kDa polypeptide comprising the TNF-homologous region of RANKL (176 amino acid residues).

  • Animal-free