Human Recombinant IL-5 (from E. coli)
Supplier: Peprotech
Certificates
About this item
IL-5 is a hematopoietic growth factor that stimulates the proliferation and activation of eosinophils. Produced by mast cells, T cells, and eosinophils, IL-5 plays an important role in inducing cell-mediated immunity against parasitic infections and certain tumors. Elevated levels of IL-5 lead to Eosinophilia, which may result in the induction of asthma and other allergic diseases. Human and murine IL-5 are cross-species reactive. Recombinant Human IL-5 is a 26.5 kDa disulfide-linked homodimeric protein containing two 116 amino acid chains.
- Animal-free
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Biological Activity:The ED50 as determined by the dose-dependent proliferation of TF-1 cells was ≤ 0.15 ng/ml, corresponding to a specific activity of ≥ 6.6 x 10^6 units/mg.
- Gene ID:3567
- Animal-Free:Y
- Protein Synonyms:Interleukin-5|EDF|TRF|BCDFII
- Protein/Peptide Name:IL-5
- Purity:98%
- Molecular Weight:26.5 kDa
- Sequence:MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
- Endotoxin Level:Low
- Formulation:Lyophilized
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together