Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Enterokinase (from CHO cells)
 :  Peprotech
 
Human Recombinant Enterokinase (from CHO cells)
 :  Peprotech

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Biological Activity:
    Sequentially cleaves carboxyl side of D-D-D-D-K.
  • Gene ID:
    5651
  • Animal-Free:
    Y
  • Protein/Peptide Name:
    Enterokinase
  • Purity:
    90%
  • Molecular Weight:
    108.7 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized
  • Shipping Temperature:
    Ambient