Anti-IgG Rabbit Polyclonal Antibody (Biotin) IGF-II
Catalog # 10775-030
Supplier: Peprotech
Specifications
- Antibody Type:Secondary
- Antigen Name:IGF-II
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:50 ug
- Cross Adsorption:No
- Form:Lyophilized
- Gene ID:3481
- Antigen Synonyms:Insulin-like Growth Factor-II, Somatamedin A
- Storage Buffer:See Product Data Sheet for more information.
- Molecular Weight:7.5 kDa
- Storage Temperature:Store at -20°C prior to reconstitution.
- Shipping Temperature:Ambient
- Immunogen:E.coli-derived Recombinant Human IGF-II (PeproTech catalog# 100-12)
- Purification:Affinity purified
- Cat. No.:10775-030
- Packaging:PP Vial
- Supplier no.:500P12BT50UG
Specifications
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-II. Anti-Human IGF-II-specific antibody was purified by affinity chromatography and then biotinylated.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human