ProNex® NGS Library Quant Kit, Promega
Catalog # 76201-682
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:ProNex® NGS Library Quant Kit
- Cat. No.:76201-682
- No. of reactions:500
Specifications
About this item
This ProNex® NGS library quant kit contains reagents to determine the concentration of next generation sequencing libraries compatible with Illumina platforms using qPCR.
- Measure intact, functional library concentration
- BRYT Green® dye-based qPCR system for maximum reproducibility
- Ideal for multiplexed pooling of Illumina libraries
Data generated using the ProNex® NGS library quant kit can be used to normalize the DNA concentration for sequencing libraries to the desired concentration prior to multiplex pooling of samples.
Delivery: Includes ProNex® NGS library Quant 2X master mix (5×1 ml), ProNex® NGS library Quant 10X primer mix (1×1 ml), ProNex® NGS Quant DNA standard (1×25 μl), ProNex® NGS library dilution buffer (2×150 ml) and nuclease-free water (2×1250 μl).
Caution: For research use only. Not for use in diagnostic procedures. Storage Temperature –30...–10 °C