ContactUsLinkComponent
Universal 2 Prong Clamp with Bosshead, Eisco Scientific
Universal 2 Prong Clamp with Bosshead, Eisco Scientific
Catalog # 470231-448
Supplier:  Wards
undefined
Universal 2 Prong Clamp with Bosshead, Eisco Scientific
Catalog # 470231-448
Supplier:  Wards
Supplier Number:  CH0688B
Specifications
  • Description:
    Universal 2 prong clamp with bosshead
  • Cat. No.:
    470231-448
Specifications
About this item
A versatile clamp to securely grip a variety of cylindrically shaped glass and labware.
  • Manufactured with stamped steel
  • Three adjustable thumb screws
  • Integrated boss head
  • Features one flat angled jaw and one V shaped jaw
  • Jaws are coated in black vinyl
  • Provides a secure grip for all cylindrical shaped objects
High quality, reliable universal steel clamp for holding and securing a variety of cylindrically shaped labware. Features include an integrated boss head, thumb screws, and vinyl coated jaws.