Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Ready to use H&E Stain Kit #1
- Size:12 lbs.
- Cat. No.:76318-762
- Application:Hematoxylin and Eosin Staining
Specifications
About this item
Validated, optimized H&E staining protocol for the Prisma automatic slide stainer.
- Patented, custom-designed bottle with ergonomic grips make easy to handle and less spills
- Each kit and component are barcoded to enable scanning of the lot numbers and expire date for documentation on Prisma Plus automatic stainer
- Biological stain commission-certified dyes are use in the manufacturing process to ensure highest quality components
- Single kit will stain up to 2,500 slides and last up to two weeks
- Color-coded labels and bottle caps which match the colors of the solution configuration of the Prisma Plus stainer and avoices filling errors
- The 750 ml fill size, ready-to-use bottles are designed to match the recommended volume for standard Prisma stainer solution reservoirs
The ready to use kit consists of 6 bottles (1 each hematoxylin and eosin, 2 each differentiator and bluing reagent) eliminating the need to measure or dilute reagents.
Shelf life: Unopened 18 months from date of manufacture, once opened 14 days.