Squishy Circuits, Standard Kit
470317-842
- Description:Squishy Circuits Standard Kit
- Environmentally Preferable:
- Cat. No.:470317-842
The standard kit includes everything you need to get started with the more advanced Squishy Circuits projects. Easy and fun!
Breakthrough your limitations by using your creativity and engineering skills to make your projects ground-breaking and one of a kind with LEDs, buzzers, and insulating dough.
: Kit includes 1×Battery holder, 1×Piezoelectric buzzer, 25×LEDs (Five each red, green, white, yellow and blue), 1 to 3.5 Oz. Container of Insulating Dough, and 3 to 3.5 Oz. Containers of Conductive Dough (in Red, Green, and Blue).