GOJO® Scrubbing Towels
Catalog # 10714-050
Supplier: Janitorial Supplies
Specifications
- Color:Silver/Yellow
- Length:12"
- Environmentally Preferable:
- Cat. No.:10714-050
- Packaging:Bucket
- Supplier no.:GOJ6396-06
- No. of towels/wipes per container:72
- Ply:2
Specifications
About this item
Dual textured with a scrubbing side for removing tough soils and a smooth side for quick dirt absorption.
- Extra-large towels for superior cleaning capacity
- Contains skin conditioners to help prevent dryness and irritation
- For use anywhere heavy duty cleaning is required; no water needed
- Fresh citrus scent
- Effective on oil, grease, paint, adhesives, tar, and asphalt
- Luxury foam handwash
Dual-textured wet towels for quick, convenient cleaning. For use on oil, grease, oil based paint, tar, adhesives, mud, clay, soil, brake dust, carbon black, shellacs, urethanes, varnish, wood stain, caulk, silicone sealers, gasket sealers, asphalt, asphalt sealers, creosote, modern printing ink