ContactUsLinkComponent
Sani-Cloth® Prime Germicidal Disposable Wipes, PDI®
Sani-Cloth® Prime Germicidal Disposable Wipes, PDI®
Catalog # 76337-090
undefined
Sani-Cloth® Prime Germicidal Disposable Wipes, PDI®
Catalog # 76337-090
Supplier Number:  PDI P25372
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Sani-Cloth® Prime Germicidal Disposable Wipe
  • Environmentally Preferable:
  • Cat. No.:
    76337-090
  • Packaging:
    Large Canister
  • W×L:
    6.0×6.75"
Specifications
About this item
Powered by ACCELOQUAT™ formulation, a next generation proprietary blend of quaternary ammonium, isopropyl alcohol (IPA), and ethanol. This unique combination delivers a true 1-minute bactericidal, fungicidal, virucidal, and tuberculocidal disinfectant. It also provides the power to destroy over 50 microorganisms, including 17 multi-drug resistant organisms (MDROs) like MRSA, CRE, and VRE.
  • Disinfection level: EPA-registered intermediate level disinfectant
  • Efficacy: Bactericidal, virucidal, fungicidal, tuberculocidal
Sani-Cloth® Prime Germicidal Disposable Wipes may be used on hard nonporous surfaces in healthcare facilities such as, bed railings, blood glucose meters (glucometers), cabinets, carts, chairs, counters, dental unit instruments trays, exam tables, gurneys, isolettes, IV poles, stethoscopes, stretchers, tables, telephones, toilet seats, diagnostic equipment, patient monitoring equipment, patient support and delivery equipment, and more. Please refer to the product/master label to get the full listing of use sites.