Specifications
- Description:Prevantics® Swab 1.0 ml, Treatment Area Covered: 2.5×2.5"
- Cat. No.:76337-046
Specifications
About this item
Prevantics® products, designed with 3.15% Chlorhexidine Gluconate (w/v) and 70% Isopropyl Alcohol (v/v) formulation, help prevent the spread of infections and help improve patient outcomes.
- No glass to break
- Pre-activated
- No waiting
- Easy to open, easy to use
- Pre-moistened
- Swab: Intuitive prep-pad design
- Swabsticks: Unique two-sided applicator for interdigital areas and skin folds
Prevantics® products are the first and only FDA approved.