ContactUsLinkComponent
Prevantics® Device Swab and Strip, PDI Healthcare
Prevantics® Device Swab and Strip, PDI Healthcare
Catalog # 76337-048
undefined
Prevantics® Device Swab and Strip, PDI Healthcare
Catalog # 76337-048
Supplier Number:  PDI B123ST
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Prevantics® Device Swab and Strip
  • Cat. No.:
    76337-048
Specifications
About this item
Disinfect needleless access sites prior to use by scrubbing the hub.
  • Confirmed efficacy at the five second scrub time and five second dry time
  • Achieved a >4.0 log10 reduction versus a leading alcohol impregnated cap against seven tested clinically relevant microorganisms known to cause CLABSIs2
The first FDA-cleared medical device featuring a 3.15% (w/v) Chlorhexidine gluconate and 70% (v/v) Isopropyl alcohol formulation.