Wipes are effective against 45 microorganisms in three minutes. Protecting staff and patients has never been easier. The alcohol-free and fragrance-free formula is ideal for use around patients, residents and staff, especially those with respiratory sensitivities.
- Short three minute contact time is effective against enveloped/non-enveloped viruses, bacteria, TB, fungi, multi-drug resistant organisms, and blood borne pathogens
- Alcohol, fragrance, bleach, acid, phenol, acetone, and ammonia-free
- Bactericidal, Tuberculocidal, Virucidal
Wipes may be used on hard, nonporous surfaces in healthcare facilities such as bed railings, blood glucose meters (glucometers), cabinets, carts, chairs, counters, dental unit instruments trays, exam tables, gurneys, isolettes, IV poles, stethoscopes, stretchers, tables, telephones, toilet seats, diagnostic equipment, patient monitoring equipment, patient support and delivery equipment, and more. Please refer to the product/master label to see a full listing of use sites.