Order Entry
United States
ContactUsLinkComponent
Rubbermaid® Commercial Brute® Container Universal Drum Dolly
Rubbermaid® Commercial Brute® Container Universal Drum Dolly
  66103-427
undefined
Rubbermaid® Commercial Brute® Container Universal Drum Dolly
  66103-427
 :  RCP2650BLA

 

  • Color:
    Black
  • Description:
    Universal Drum Dolly
  • Dimensions:
    61.9Ø×18.1H cm (243/8×71/8")
  • Material:
    Plastic
  • Environmentally Preferable:
  • Cat. No.:
    66103-427
  • For use with:
    Brute® 55 gal. Round Containers
  • Casters:
    4 swivel, TPR
  • Supplier no.:
    RCP 2650 BLA
  • Caster diameter:
    3"

 

 

The Rubbermaid® Commercial universal drum dolly cart quickly and easily transports waste containers up to 500 lbs. throughout your facility.

  • Easily transports waste containers up to 500 lbs.
  • Makes waste collection more efficient, saves time, and reduces worker strain
  • Resists rust, chipping, and denting
  • Heavy-duty swivel casters enhance mobility
  • Drop-in connection

Easy to maneuver, this cart dolly makes waste collection more efficient, saves time, and reduces worker strain from heavy loads. It holds plastic containers, as well as 55 gal. drums. The dependable, heavy-duty swivel casters provide enhanced mobility through busy work areas or tight spaces. For use with the Rubbermaid® Commercial waste receptacles.