Specifications
- Description:CHOgro® Transfection and Titer Enhancer Kit
- Size:1 and 20 ml
- Cat. No.:76325-206
Specifications
About this item
The most advanced system for transient transfection and protein production in suspension CHO cells.
- High yield - Reach higher antibody titers in seven days which is faster than the ExpiCHO expression system
- Simple workflow - Same day transfection, enhanced addition and temperature shift
- Worry free - No commercial license required
The CHOgro® high yield expression system is an optimized platform for transient, high titer protein production in suspension CHO derived cells. This system consists of CHOgro® expression medium, L-Glutamine and Poloxamer 188 medium supplements, CHOgro® complex formation solution, TransIT-PRO® transfection reagent and CHOgro® titer enhancer. CHOgro® expression medium is a chemically defined, hydrolysate free and animal origin free medium manufactured using cGMP compliant processes in an ISO-compliant facility. CHOgro® medium is formulated to support high efficiency transfection and provide high density cell growth. In addition, many suspension CHO cells (e.g. Freestyle™ CHO-S and ExpiCHO-S™) can easily grow in CHOgro® medium with minimal adaptation.
TransIT-PRO® transfection reagent was developed by empirically testing proprietary lipid and polymer libraries for high transfection performance in suspension CHO and 293 cell types. The CHOgro® titer enhancer was identified and developed for use with the CHOgro® high yield expression system to modulate cellular processes and increase protein titers.
Both the TransIT-PRO® reagent and CHOgro® titer enhancer are free of animal derived components. The combination of CHOgro® expression medium, TransIT®-PRO transfection reagent and the CHOgro® titer enhancer enable robust cell growth and high efficiency transfection that streamlines the transient protein expression process.