Order Entry
Puerto Rico
ContactUsLinkComponent
Eosin Y (yellowish) alcoholic solution, Surgipath® ready-to-use, used in nuclear staining
Eosin Y (yellowish) alcoholic solution, Surgipath® ready-to-use, used in nuclear staining
Catalog # 10015-072
Supplier:  Leica Biosystems
CAS Number:  
undefined
Eosin Y (yellowish) alcoholic solution, Surgipath® ready-to-use, used in nuclear staining
Catalog # 10015-072
Supplier:  Leica Biosystems
Supplier Number:  3801600
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 gal.
  • Cat. No.:
    10015-072
  • Supplier no.:
    3801600

Specifications

About this item

 :
 :691.86 g/mol
CAS Number:17372-87-1
Formula:C₂₀H₆Br₄Na₂O₅
MDL Number:MFCD00005040