Order Entry
Puerto Rico
ContactUsLinkComponent
Hematoxylin solution (Harris) used in nuclear staining mercury-free
Hematoxylin solution (Harris) used in nuclear staining mercury-free
Catalog # 10015-074
Supplier:  Leica Biosystems
CAS Number:  
undefined
Hematoxylin solution (Harris) used in nuclear staining mercury-free
Catalog # 10015-074
Supplier:  Leica Biosystems
Supplier Number:  3801560
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 gal.
  • Cat. No.:
    10015-074
  • Supplier no.:
    3801560

Specifications

About this item

 :
 :302.28 g/mol
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111