ContactUsLinkComponent
SecureSeal™ Adhesive Film for Microplates, Simport Scientific
SecureSeal™ Adhesive Film for Microplates, Simport Scientific
Catalog # 14211-710
Supplier:  Simport Scientific
undefined
SecureSeal™ Adhesive Film for Microplates, Simport Scientific
Catalog # 14211-710
Supplier:  Simport Scientific
Supplier Number:  T329-3
Specifications
  • Sterility:
    Non sterile
  • Description:
    SecureSeal™ Adhesive Film for Microplates
  • Cat. No.:
    14211-710
  • Supplier no.:
    T329-3
Specifications
About this item
This adhesive sealing films reduce sample-to-sample or well-to-well contamination and or spill over.
  • Compatible with almost all microplate procedures
  • RNase, DNase, DNA-free
  • DMSO resistant
  • Temperature range: –40...+120 °C
The polyester based film with acrylic adhesive is inert and thus compatible with almost all microplate procedures. SecureSeal™ is less than .001% moisture permeable under high humidity conditions and less than .01% oxygen permeable.
Packaging: Each package contains 100 sheets.