Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DPA/Terbium set
DPA/Terbium set for membrane fusion assay
DPA/Terbium set for membrane fusion assay
DPA/Terbium set for membrane fusion assay
Catalog # 89139-510
Supplier:
Biotium
CAS Number:
Expand image
DPA/Terbium set for membrane fusion assay
Catalog # 89139-510
Supplier:
Biotium
Supplier Number:
80104
CAS Number:
Specifications
Size:
1 set
Cat. No.:
89139-510
Supplier no.:
80104
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
:
499-83-2
Product Details