Order Entry
Puerto Rico
ContactUsLinkComponent
Biotin-X-DHPE membrane stain, for histology
Biotin-X-DHPE membrane stain, for histology
Catalog # 89139-322
Supplier:  Biotium
CAS Number:  
Biotin-X-DHPE membrane stain, for histology
Catalog # 89139-322
Supplier:  Biotium
Supplier Number:  60023
CAS Number:  

Specifications

  • Size:
    5 mg
  • Cat. No.:
    89139-322
  • Supplier no.:
    60023

Specifications

About this item

 :
 :1132.60 g/mol
Storage Temperature:Freezer
Formula:C₅₉H₁₁₄N₅O₁₁PS