ChemMax® 2 Chemical Protective Aprons, Lakeland Industries
Supplier: Lakeland
Certificates
About this item
Chemical protective aprons are made with the proven protection of Saranex® 23P barrier film for protection in laboratory, chemical handling, and environmental clean up applications.
- Long sleeve
- Elastic wrist closures
These aprons provide the protection needed for chemical mixing and splash and hazardous materials remediation.
Specifications
- Catalog No:
- 76332-666
- Garment application:
- Laboratory