Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PD-1/PD-L1 Inhibitor 2
PD-1/PD-L1 Inhibitor 2 ≥98% (by HPLC)
PD-1/PD-L1 Inhibitor 2 ≥98% (by HPLC)
PD-1/PD-L1 Inhibitor 2 ≥98% (by HPLC)
Supplier:
Avantor
Certificates
Expand image
PD-1/PD-L1 Inhibitor 2 ≥98% (by HPLC)
Supplier:
Avantor
About this item
CAS Number:
1675203-84-5
:
Formula:
C₂₅H₂₉N₃O₃
MDL Number:
MFCD29905455
:
419.52 g/mol
Storage Temperature:
Freezer
Related Information
Documentation
Product specification
Spec_PD-1 or PD-L1 Inhibitor 2.pdf
Documentation