Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PX-478 hydrochloride
PX-478 hydrochloride ≥98% (by HPLC)
PX-478 hydrochloride ≥98% (by HPLC)
PX-478 hydrochloride ≥98% (by HPLC)
Supplier:
Avantor
Certificates
Expand image
PX-478 hydrochloride ≥98% (by HPLC)
Supplier:
Avantor
About this item
CAS Number:
685898-44-6
:
Formula:
C₁₃H₂₀Cl₄N₂O
MDL Number:
MFCD11840749
:
394.11 g/mol
Storage Temperature:
Freezer
Related Information
Documentation
Product specification
Spec_PX-478 Hydrochloride.pdf
Documentation