Biohazard Spill Kit, Bucket, Spill Magic, Acme United
Catalog # 76327-120
Supplier: First Aid Only
Specifications
- Description:Biohazard Spill Clean Up
- Height:9"
- Length:10.5"
- Size:4/pack
- Cat. No.:76327-120
- Packaging:Bucket
- Weight:12 oz.
- Width:10"
Specifications
About this item
Spill magic single use biohazard cleanup kit safely removes blood and bodily fluids.
- 12 oz. bag of all-purpose spill clean up to quickly absorb and remove spill
- Personal protective equipment components of the kit help minimize exposure of personnel to potentially harmful substances
- Safely clean up potentially infectious bodily fluids such as vomit, spit, blood, or urine with tools and disposal bags
- Disposable single use kit to prevent crosscontamination
The disposable kit includes 12 oz. Spill Magic Absorbent, plastic bag, goggles, gloves, brush and dustpan, chemical surface wipe, anti-microbial towelette and I.D. tag.
Accessories information: Tools needed to clean up spill included.
Caution: Used Spill Magic must be disposed of in accordance with any federal, state or local laws and regulations governing the absorbed liquid or material. Do not use to absorb hydrofluoric acid. Do not pour down drain.