Orders LinkContactUsLinkComponent
Loopster Digital Rotating Shaker, IKA
Loopster Digital Rotating Shaker, 100/240 V, 50/60 Hz, 300×300×360 mm, 0 - 80 rpm, 0 - 90°, 3 kg
Catalog # 76329-832
Irregular Voltage
Supplier:   IKA Works
 :  0004016000
Loopster Digital Rotating Shaker, IKA
Catalog # 76329-832
Supplier:  IKA Works
Irregular Voltage
Loopster Digital Rotating Shaker, IKA
Loopster Digital Rotating Shaker, IKA
Loopster Digital Rotating Shaker, 100/240 V, 50/60 Hz, 300×300×360 mm, 0 - 80 rpm, 0 - 90°, 3 kg
Price
$1,906.62
Log in for availability
Each
Product Details & Documents
Digital rotator for smooth but effective mixing with adjustable speed. Ideal for biological samples, particularly blood samples.
  • Usable at 4 to 50 °C in incubators
  • Freely adjustable rotary angle
  • Suitable for continuous operation
  • Error code display
  • Easy operation by foil keypad
  • Timer: Countdown, adjustable from 1 second to 5999 minutes
Suitable for separating and mixing processes of powdered and liquid samples in Eppendorf tubes and Greiner tubes up to 50 ml. The easy removable attachments allow simple and quick cleaning of the unit of spilled samples.
 : IP 21 protection class according to DIN EN 60529.
Specifications
  • Description:
    Loopster Digital Rotating Shaker
  • W×D×H:
    300×300×360 mm
  • Electrical:
    100/240 V, 50/60 Hz
  • Speed range:
    0 - 80 rpm
  • Tilt angle:
    0 - 90°
  • Weight:
    3 kg
  • Shaking motion:
    Rotating
  • Cat. No.:
    76329-832
  • Timer:
    1 Second to 5999 Minutes
  • Max. Load:
    2 kg
  • Model:
    Loopster Digital
  • Display:
    7 Segment LED
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • DS 6.2, 12 spare clips for Loopsters with Ø 12 mm
    DS 6.2, 12 spare clips for Loopsters with Ø 12 mm
    76329-666
    Log in for availability
    Pack of 12 - $26.24
Product Information
  • Catalog #
    Availability
    Price
  • DS 6.3, 12 spare clips for Loopsters with Ø 16 mm
    DS 6.3, 12 spare clips for Loopsters with Ø 16 mm
    76329-668
    Log in for availability
    Pack of 12 - $41.44
Product Information
  • Catalog #
    Availability
    Price
  • DS 6.4, 12 spare clips for Loopsters with Ø 25 mm
    DS 6.4, 12 spare clips for Loopsters with Ø 25 mm
    76329-670
    Log in for availability
    Pack of 12 - $42.97
Product Information
  • Catalog #
    Availability
    Price
  • DS 6.5, 12 spare clips for Loopsters with Ø 28 mm
    DS 6.5, 12 spare clips for Loopsters with Ø 28 mm
    76329-672
    Log in for availability
    Pack of 12 - $36.87
Product Information
  • Catalog #
    Availability
    Price
  • DS 6 Universal disc for Loopster, flexibly equippable with spare clips (available separately)
    DS 6 Universal disc for Loopster, flexibly equippable with spare clips (available separately)
    76329-678
    Log in for availability
    Each - $397.46
Product Information
  • Catalog #
    Availability
    Price
  • DS 2 Tube attachment for Loopster for 24×5 ml tubes with Ø 12 mm
    DS 2 Tube attachment for Loopster for 24×5 ml tubes with Ø 12 mm
    76329-782
    Log in for availability
    Each - $476.92
Product Information
  • Catalog #
    Availability
    Price
  • DS 1 Tube attachment for Loopster for 48×1.5 ml tubes with Ø 10 mm
    DS 1 Tube attachment for Loopster for 48×1.5 ml tubes with Ø 10 mm
    76329-780
    Log in for availability
    Each - $476.92
Product Information
  • Catalog #
    Availability
    Price
  • DS 3 Tube attachment for Loopster for 20×15 ml tubes with Ø 16 mm
    DS 3 Tube attachment for Loopster for 20×15 ml tubes with Ø 16 mm
    76329-784
    Log in for availability
    Each - $482.00
Product Information
  • Catalog #
    Availability
    Price
  • DS 4 Tube attachment for Loopster for 12×30 ml tubes with Ø 25 mm
    DS 4 Tube attachment for Loopster for 12×30 ml tubes with Ø 25 mm
    76329-786
    Log in for availability
    Each - $476.92
Product Information
  • Catalog #
    Availability
    Price
  • DS 5 Tube attachment for Loopster for 12×50 ml tubes with Ø 28 mm
    DS 5 Tube attachment for Loopster for 12×50 ml tubes with Ø 28 mm
    76329-788
    Log in for availability
    Each - $482.00
Product Information
  • Catalog #
    Availability
    Price
  • DS 6.1, 12 spare clips for Loopsters with Ø 10 mm
    DS 6.1, 12 spare clips for Loopsters with Ø 10 mm
    76329-964
    Log in for availability
    Pack of 12 - $25.37