Biohazard Cleanup Kit, Spill Magic, Acme United
76327-118
- Description:Biohazard Spill Clean Up
- Height:10.5"
- Length:18.5"
- Size:4/pack
- Cat. No.:76327-118
- Packaging:BAGA
- Weight:8 oz.
- Width:13"
Spill magic single use biohazard cleanup kit safely removes blood and bodily fluids.
- Biohazard clean Up kit contents meet OSHA standards
- Personal protective equipment components of the kit help minimize exposure of personnel to potentially harmful substances
- Safely clean up potentially infectious bodily fluids such as vomit, spit, blood, or urine with tools and disposal bags
- Disposable single use kit to prevent crosscontamination
The disposable kit includes 8 oz. spill magic absorbent, plastic bag, apron, gloves face mask shield, brush and dustpan, chemical surface wipe, anti-microbial towelette, and biohazard bag with I.D. tag.
: Tools needed to clean up spill included.
: Used Spill Magic must be disposed of in accordance with any federal, state or local laws and regulations governing the absorbed liquid or material. Do not use to absorb hydrofluoric acid. Do not pour down drain.