Order Entry
Puerto Rico
ContactUsLinkComponent
All-Purpose Spill Clean Up Bag, Spill Magic, Acme United
All-Purpose Spill Clean Up Bag, Spill Magic, Acme United
Catalog # 76327-186
Supplier:  First Aid Only
undefined
All-Purpose Spill Clean Up Bag, Spill Magic, Acme United
Catalog # 76327-186
Supplier:  First Aid Only
Supplier Number:  SM12-001

Specifications

  • Description:
    Absorbent powder
  • Height:
    8.5"
  • Length:
    11.25"
  • Size:
    12/pack
  • Cat. No.:
    76327-186
  • Packaging:
    BAGA
  • Form:
    Powder
  • Weight:
    12 oz.
  • Width:
    2"

Specifications

About this item

Spill magic lightweight, high performance, all-purpose absorbent powder removes a liquid or semi-liquid spill of any source or viscosity almost instantly.

  • 12 oz. bag of absorbent powder
  • Quick and easy to use
  • Non-toxic
  • Absorbs spill instantly leaving the surface clean and dry
  • For any spill (except Hydrofluoric Acid)
  • Single use plastic bag for one time clean up

Spill magic is a quick clean up solution that leaves the surface dry and residue free.

Save time, money and reduce slip and fall instances. The powder is ideal for cleaning industrial, food or biohazard spills.

Accessories information: Accessories not included.
Caution: Used spill magic must be disposed of in accordance with any federal, state or local laws and regulations governing the absorbed liquid or material. Do not use to absorb hydrofluoric acid. Do not pour down drain.