Single-Wire Adjustable Metal Wrist Strap, SCS, Desco Industries
89024-454
: Desco
- Description:Single-Wire Adjustable Metal Wrist Strap
- Environmentally Preferable:
- Cat. No.:89024-454
- Supplier no.:AMWS
Made from stainless steel, the durable, premium performance metal wristband creates a reliable electrical contact to the user’s wrist.
- Flex-metal style band uses adjustable links to assure a secure and comfortable fit
The retractable coil cord terminated with a banana plug on one end and connects to the wristband via a 4 mm post snap. It contains a 1 megohm resistor and includes alligator clip.
: Meets requirements of ANSI/ESD S20.20 and Wrist Strap standard ANSI/ESD S1.1.