GTS Rx Overpack Boxes, Cold Chain Technologies
Supplier: Cold Chain Technologies
Certificates
About this item
Overpack boxes are designed for use with GTS Rx shippers, offering four extra inches of height on top of the shipper for accessories that do not have thermal requirements.
- Made from environmentally safe and reusable materials
- Designed for research and development shipments
Recommended uses are specialty and mail-order pharmacies and drug sampling programs. Overpack Boxes are compatible with the GTS Rx shipper family line of parcel containers.