Vesta-Syde® SQ 128 Ready-To-Use (RTU) Disinfectants
Supplier: Steris
Certificates
About this item
STERIS' Vesta-Syde® SQ 128 ready to use quaternary ammonium disinfectant is specifically formulated for disinfection of hard surfaces. The Ready to Use solution can be applied manually or sprayed.
- Ease of use - Eliminates the need to validate dilution preparation procedures by providing a consistent dilution every time
- Convenience - Same key ingredients as Vesta-Syde® SQ Disinfectant concentrate to simplify validation requirements
- Productivity - Low residue formulation to maintain visibly clean surfaces with minimal rinsing
- Efficacy - Microbial control against bacteria, fungi, and viruses using AOAC and EN protocols
- Availability - Active ingredients notified according to the Biocide Product Directive and registered with the US EPA
- Assurance - Manufactured in accordance with the cGMP with complete lot traceability
STERIS’ Vesta-Syde® SQ 128 ready to use quaternary ammonium disinfectant is the non sterile version of our phosphate-free one-step cleaner and disinfectant formulated for use on hard, non-porous surfaces.
Made with same main ingredients as the Vesta-Syde® SQ disinfectant concentrate, Vesta-Syde® SQ RTU disinfectants kill a broad spectrum of common bacteria and fungi. At a use-dilution rate of 0.8% v/v (1:128), Vesta-Syde® SQ 128 ready to use disinfectant requires no mixing or activation for convenient and consistent use.