ContactUsLinkComponent
Versaflo™ PAPR Assembly TR-307N+, with Easy Clean Belt and High Capacity Battery, 3M™
Versaflo™ PAPR Assembly TR-307N+, with Easy Clean Belt and High Capacity Battery, 3M™
Catalog # 76220-726
Supplier:  3M
undefined
Versaflo™ PAPR Assembly TR-307N+, with Easy Clean Belt and High Capacity Battery, 3M™
Catalog # 76220-726
Supplier:  3M
Supplier Number:  7100153810
Specifications
  • Description:
    Versaflo™ PAPR Assembly TR-307N+, with Easy Clean Belt and High Capacity Battery
  • Cat. No.:
    76220-726
Specifications
About this item
Designed to provide protection against particulates, the Versaflo™ PAPR Assembly TR-307N+ combines performance, protection, and functionality with a slim, lightweight, balanced design for comfort.
  • Two selectable flow rate options (6.5 and 7.2 CFM) offer more options and comfort to the user
  • Easy-to-use and versatile PAPR system for particulate environments
  • High efficiency particulate filter
  • High efficiency battery allows work in a remote location for extended periods of time
Two types of alarms (one audible and one visual) alert user to either a low battery or low flow, such as when a particle filter is fully loaded. LED lights on the battery indicate charge status.
High capacity lithium ion rechargeable battery provides a life of 10 to 12 hours. Belt-mounted tube type. Use in machine operation and power handling applications.
Certifications: NIOSH approved.
Delivery: The Versaflo™ TR-307N+ PAPR assembly includes a TR-302N+ PAPR unit, TR-371+ filter cover, TR-327 easy clean belt, TR-332 battery, TR-341N battery charger kit, TR-3712N HE filter, and a TR-3600 Pre-Filter.