DURAN® GLS 80® Double Walled Bottle, Wide Mouth, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
The DURAN® double walled, wide mouth bottles GLS 80® incorporate an integral jacket that isolates the contents from the external environment.
- Autoclavable at 121 °C
- Offers a sealable and more flexible alternative to open topped jacketed beakers
Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN® bottle.
Certifications: USP standards.