Oakton® pH and ORP Electrodes, Cole-Parmer
Supplier: Antyila Scientific
Certificates
About this item
Reliable and cost effective probes.
- Won't break - epoxy electrodes resist impact and are extremely durable
- Glass electrodes resist aggressive chemicals
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with the sample.
Choose junction and tip style based on your application. All electrodes feature Ag/AgCl reference cells.
Specifications
- Catalog No:
- 76326-466
- 76326-442
- 76326-464
- 76326-462
- 76326-458
- 76326-456
- 76326-454
- 76326-452
- 76326-450
- 76326-496
- 76326-448
- 76326-498
- 76326-500
- 76326-446
- Connection:
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- BNC (pH/ORP)
- Cable Length:
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')
- 1 m (3')