Order Entry
Puerto Rico
ContactUsLinkComponent
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (by HPLC) mitochondria dye, suitable for fluorescence
Irritant
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (by HPLC) mitochondria dye, suitable for fluorescence
Catalog # 89139-392
Supplier:  Biotium
CAS Number:  
Irritant
Tetramethylrhodamine methyl ester perchlorate (TMRM) ≥95% (by HPLC) mitochondria dye, suitable for fluorescence
Catalog # 89139-392
Supplier:  Biotium
Supplier Number:  70017
CAS Number:  

Specifications

  • Size:
    25 mg
  • Cat. No.:
    89139-392
  • Supplier no.:
    70017

Specifications

About this item

 :
 :500.93 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:115532-50-8
Formula:C₂₅H₂₅N₂O₃·ClO₄
MDL Number:MFCD00274460