


Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:51811-82-6
:
MDL Number:MFCD00081642
Boiling Pt:>65 °C (1013 hPa)
Density:0.99 g/cm3 (20 °C)
Flash Pt:18 °C
Storage Temperature:Ambient
UN:1230
ADR:3 (6.1),II
:M-Clarity™ quality level = MQ300
:GHS06
:GHS02
:GHS08
:1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Suitability for microscopy:passes test
- Erythrocytes:reddish-brownish
- Nuclei:violet
- Eosinophilic granules:reddish to red-brown
- Neutrophilic granules:light violet
- Lymphocyte cytoplasm:Blue
- Monocyte plasma:grey-blue
- Thrombocytes:violet
- Basophilic granules:dark violet
- Blood parasite nuclei:light red
- Nuclei (Tissue section):blue to dark blue
- Cells:blue to dark blue
- Collagen:pale blue
- Osteoid:pale blue
- Eosinophilic granula (Tissue section):red
- Acidic mucopolysaccharides:reddish-violet
- Mast cell granules:reddish-violet
- Cartilage matrix:reddish-violet
- Acidophilic materials:orange-red
- Helicobacter pylori:blue to dark blue