Specifications
- Description:Steri-Dot® Indicators
- Environmentally Preferable:
- Cat. No.:76238-042
Specifications
About this item
Steri-Dot® indicators provide a fast positive visual check for processed packages sterilized in ethylene oxide.
- Self-adhesive ethylene oxide process indicator
- Permanent Red to Green Color change on exposure to EO gas
- Decipher between processed and unprocessed packages
These indicators offer a fast positive visual check for processed ethylene oxide sterilized items. Indicator changes from red to green after exposure to EO sterilization cycle. As an additional safeguard, in the absence of adequate humidity, the color change to green will not occur.