Order Entry
United States
ContactUsLinkComponent
ARM-D Kit, β-Lactamase, Steck
ARM-D Kit, β-Lactamase, Steck
Catalog # 76318-696
CAS Number:  
undefined
ARM-D Kit, β-Lactamase, Steck
Catalog # 76318-696
Supplier Number:  250045
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ARM-D Kit, β-Lactamase (RUO)
  • Cat. No.:
    76318-696
  • No. of reactions:
    100

Specifications

About this item

The Streck ARM-D Kit, β-Lactamase, is a comprehensive, TaqMan based method for the detection of antibiotic resistance gene families in Gram Negative Bacteria, including the five CRE's described by the CDC as urgent public health threats.

  • Detects over 450 gene variants
  • Dectects the five major CRE resistance families
  • Dectects ESBL's and ampC resistance markers
  • Understand resistance mechanisms prior to obtaining susceptibility results
  • Works on a large variety of instrumentation