ContactUsLinkComponent
ARM-D Kit, ampC (RUO), Streck
  76318-694
 :  
undefined
ARM-D Kit, ampC (RUO), Streck
  76318-694
 :  250044
 :  
  • Description:
    ARM-D Kit, AmpC (RUO)
  • Cat. No.:
    76318-694
  • No. of reactions:
    100
The Streck ARM-D Kit, AmpC, is a comprehensive, TaqMan based method for the detection of plasmid based antibiotic resistance gene families in Gram Negative Bacteria.
  • Dectects the six major ampC resistance families
  • The only commercially available multiplex kit to identify plasmid-mediated AmpCs
  • Understand resistance mechanisms prior to obtaining susceptibility results
  • Works on a large variety of instrumentation