Bioruptor® Plus Microtubes, Diagenode
Catalog # MSPP-C30010013
Supplier: Diagenode
Specifications
- Description:Bioruptor® Plus Microtubes
- Size:0.5 ml
- Cat. No.:MSPP-C30010013
- Packaging:500/Pack
Specifications
About this item
Bioruptor® microtubes are designed for DNA and chromatin shearing and strongly improve sonication efficiency.
- Recommended sample volume: 50 to100 μl
These tubes have been validated to be used with Bioruptor® Standard and Bioruptor® Plus.