ContactUsLinkComponent
Jess or Wes Separation Module, Protein Simple
Jess or Wes Separation Module, Protein Simple
Catalog # MSPP-SMW002
Supplier:  PROTEIN SIMPLE MS
CAS Number:  
Jess or Wes Separation Module, Protein Simple
Catalog # MSPP-SMW002
Supplier:  PROTEIN SIMPLE MS
Supplier Number:  SM-W002
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Jess or Wes Separation Module
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-SMW002
Specifications
About this item
Reagents for 104 data points to use with a detection module.
It includes two parts: Separation box with 8 capillary Cartridges (13 capillaries per cartridge), 8 Jess/Wes 12 to 230 kDa pre-filled Plates (pre-filled with separation matrix 2, stacking matrix 2, split running buffer 2 and matrix removal buffer), sample buffer, wash buffer, plus standard pack 1 clamshell with 8 ea. DTT, FL standard master mix with 29 kDa system control protein.