Order Entry
Puerto Rico
ContactUsLinkComponent
DNA Detection Kit in Tubes, Cygnus Technologies
DNA Detection Kit in Tubes, Cygnus Technologies
Catalog # 102650-482
Supplier:  Cygnus
CAS Number:  
DNA Detection Kit in Tubes, Cygnus Technologies
Catalog # 102650-482
Supplier:  Cygnus
Supplier Number:  D410T
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    E. coli Host Cell DNA Detection Kit in Tubes
  • Cat. No.:
    102650-482
  • Supplier no.:
    D410T

Specifications

About this item

E. coli host cell DNA detection kit method utilizes a 2 ml microfuge tube format to measure residual E. coli host cell DNA. The kit includes proprietary extraction procedure, which isolates residual DNA without the use of iodine or glycogen.

  • Detection method: PicoGreen®
  • Storage: 2 to 8 °C (all kit reagents)
  • Species group: Bacterial

The superior co-precipitating reagents do not interfere with qPCR and ensure that pellets do not become dislodged during the extraction procedure. This eliminates the need to centrifuge samples again as well as the need to retest or repeat a run. Cell line-specific E. coli DNA calibrators are included, and upon completion of extraction, samples are reacted with PicoGreen® solution. Time to completion is under 2 hours. Each kit supplies enough reagent for the analysis of 50 samples in duplicate.

Delivery: Kit includes DNA sample diluent, DNA precipitation buffer, microfuge tubes, DNA extraction buffer for tubes, assay plate with plate sealing foil, E. coli, DNA standards set, A-H (1 ml), proteinase K, DNA wash buffer and Quant-iT™ PicoGreen® dsDNA reagent solution.