Order Entry
Puerto Rico
ContactUsLinkComponent
Methyl Cellulose Kit, 1%, Protein Simple
Methyl Cellulose Kit, 1%, Protein Simple
Catalog # MSPP-101876
Supplier:  PROTEIN SIMPLE MS
CAS Number:  
Methyl Cellulose Kit, 1%, Protein Simple
Catalog # MSPP-101876
Supplier:  PROTEIN SIMPLE MS
Supplier Number:  101876
CAS Number:  

Specifications

  • Description:
    Methyl Cellulose Kit, 1%
  • Cat. No.:
    MSPP-101876

Specifications

About this item

Methyl cellulose, 1% is needed to maintain the viscosity of samples on the FC cartridge only.

Delivery: Kit includes a 100 ml bottle.