Order Entry
Puerto Rico
ContactUsLinkComponent
Rabbit Lactate Dehydrogenase (from Muscle)
Rabbit Lactate Dehydrogenase (from Muscle)
Catalog # 80601-264
Supplier:  MilliporeSigma
CAS Number:  
Rabbit Lactate Dehydrogenase (from Muscle)
Catalog # 80601-264
Supplier:  MilliporeSigma
Supplier Number:  427217-25KU
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic ampoule
  • Source:
    Muscle
  • Species:
    Rabbit
  • Size:
    25 KU
  • Storage Conditions:
    Store at –20 °C
  • Enzyme Name:
    Lactate Dehydrogenase
  • Cat. No.:
    80601-264
  • Supplier no.:
    427217-25KU

Specifications

About this item

Native lactate dehydrogenase from rabbit muscle. Catalyzes the reversible conversion of pyruvate to lactate. The overall equilibrium of this reaction favors lactate formation.

One unit is defined as the amount of enzyme that will convert 1.0 µmol pyruvate to L-lactate per min in 100 mM phosphate buffer at 30 °C, pH 7.0.