L(+)-Cysteine free base ≥98% (by titrimetric analysis), Calbiochem®, Millipore®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:52-90-4
:
Formula:HSCH₂CH(NH₂)CO₂H
MDL Number:MFCD00064306
:121.16 g/mol
Merck Index:14,02781
:Packaged under inert gas.
Storage Temperature:Ambient
Specification Test Results
- Purity (by Titration, Perchloric Acid):≥98%
- Iron:≤10 ppm
- Endotoxin:≤24 EU/g
- Total microorganisms:≤100 CFU/g
- Heavy metals:≤0.001% (as Pb)